Tisk…

Anti-PLC gamma 2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3587R-100EA 6910 CZK
BIOV3587R-100
Anti-PLC gamma 2 Rabbit Polyclonal Antibody
Protilátky
PLC (Phosphoinositide-specific phospholipase C) plays a significant role in transmembrane signaling. Four members of PLCs have been identified: PLCβ, PLCg, PLCδ, and PLCe. In response to extracellular stimuli (e.g., hormone, growth factors, neurotransmitters), PLC hydrolizes phosphatidylinositol 4,5-biphosphate (PIP2) into two secondary messengers: inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG;PLCg2 is engaged in antigen-dependent signaling in B-cells and collagen-dependent signaling in platelets.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PLC gamma 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR