Tisk…

Anti-Cathepsin K Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3588-100EA 8740 CZK
BIOV3588-100
Anti-Cathepsin K Rabbit Polyclonal Antibody
Protilátky
Cathepsin family of proteases contains several diverse classes of enzymes. The cysteine protease class comprises cathepsin B, H, K, L, O, and S. The aspartyl protease class contains cathepsin D and E. Cathepsin G belongs to the serine protease class. Cathepsins are involved in various cellular events such as peptide biosynthesis, protein degradation, and apoptosis. Cathepsin K expression is highest in bone, cartilage, and skeletal muscle.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunoprecipitation, and Immunohistochemistry

Type: Primary
Antigen: Cathepsin K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Rabbit, Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR