Tisk…

Anti-Galectin-3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3591-100EA 6910 CZK
BIOV3591-100
Anti-Galectin-3 Rabbit Polyclonal Antibody
Protilátky
Galectins are a family of soluble β-galactoside-binding animal lectins that modulate cell-to-cell and cell-to-ECM interactions, as well as apoptosis. Galectin-3 encodes a 30-35 kDa protein containing a single carbohydrate binding domain, which binds galactose-containing glycoconjμgates. Upregulated expression of Galectin-3 is involved in cancer progression and metastasis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Galectin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR