Tisk…

Anti-Dio-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3592-100EA 6910 CZK
BIOV3592-100
Anti-Dio-1 Rabbit Polyclonal Antibody
Protilátky
Apoptosis plays an important role in many cellular processes. Dio-1 (Death Inducer-O bliterator-1), a putative transcription factor, has been indicated to function in the onset of apoptosis. Dio-1 expression is upregulated when an apoptosis is detected. It translocates to the nucleus, and activates apoptosis during limb development. Dio-1 expression can be suppressed by caspase inhibitors and Bcl-2 expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Dio-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR