Tisk…

Anti-PLTP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3595-100EA 7190 CZK
BIOV3595-100
Anti-PLTP Rabbit Polyclonal Antibody
Protilátky
Plasma phospholipid transfer protein (PLTP) plays a central role in regulating plasma HDL levels and distribution of HDL subpopulations. This regulation is evidenced in vivo by markedly reduced plasma HDL levels in PLTP gene knock-out mice. PLTP also facilitates free cholesterol influx into HDL.

Recommended Dilutions: Western Blot: 1:200-1000 dilutions; in Immunohistochemistry: 1:50 dilutions

Type: Primary
Antigen: PLTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR