Tisk…

Anti-Hsc70 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3095-100EA 7900 CZK
BIOV3095-100
Anti-Hsc70 Rabbit Polyclonal Antibody
Protilátky
The 70 kDa heat shock cognate protein, Hsc70, is part of the Hsp70 family. The family includes both cognate members and highly inducible isoforms. Members of the Hsp70 family are molecular chaperones which are involved in many cellular functions such as protein folding, transport, maturation and degradation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunohistochemistry

Type: Primary
Antigen: Hsc70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Rabbit, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR