Tisk…

Anti-SIRT7 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3099-100EA 7780 CZK
BIOV3099-100
Anti-SIRT7 Rabbit Polyclonal Antibody
SIRT7 is a member of the sirtuin family of proteins (Sir2-like proteins;All Sir2-like proteins are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins are not fully understood yet. Recent studies sμggested that bacterial, yeast and mammalian sirtuins are able to metabolize NAD and may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity.

  • Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: SIRT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR