Tisk…

Anti-MAPKAPK-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3100-100EA 7470 CZK
BIOV3100-100
Anti-MAPKAPK-2 Rabbit Polyclonal Antibody
Protilátky
MAP kinase-activated protein kinase-2 (MAPKAPK-2), also known as p45 hsp27 kinase, is a 45-54 kDa serine-threonine protein kinase that contains a proline rich sequence and two putative SH3-binding sites. MAPKAPK-2 is activated in response to stress, IL-1 and TNF, possibly catalyzed by p38/Hog-dependent phosphorylation. One of the major substrates of MAPKAPK-2 is hsp27, which stimulates actin polymerization in order to facilitate recovery from destruction of cytoskeleton during cellular stresses. Two isoforms are produced due to alternative splicing of the same gene which differs in their C-terminals. This antibody recognizes both isoforms.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: MAPKAPK-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR