Tisk…

Anti-Jagged 1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3101-100EA 7780 CZK
BIOV3101-100
Anti-Jagged 1 Rabbit Polyclonal Antibody
Jagged belongs to the Delta­Serrate­Lag­2 (DSL) family of ligands that activate LIN12/Notch proteins. Ligands for Notch include Jagged1, Jagged2 and Delta. Jagged 1is involved in mammalian cardiovascular development and in cell-fate decisions during hematopoiesis. Mutation in Jagged1 is associated with an autosomal dominant disorder known as Alagille syndrome and Familial Tetralogy of Fallot a form of complex congenital heart disease.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Jagged 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR