Tisk…

Anti-Amylase pancreatic Mouse Monoclonal Antibody [clone: RPA-B5]

Dodavatel: BioVision
BIOV3102-100EA 7470 CZK
BIOV3102-100
Anti-Amylase pancreatic Mouse Monoclonal Antibody [clone: RPA-B5]
Protilátky
Pancreatic amylase is a digestive enzyme secreted by pancreas into the intestine. It consists of a single polypeptide chain with a molecular weight of 54 kDa. Measurement of amylase concentrations in serum is a diagnostic test for various pancreatic disorders.

Recommended Dilutions: indirect ELISA and WB.

Type: Primary
Antigen: Amylase pancreatic
Clonality: Monoclonal
Clone: RPA-B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR