Tisk…

Anti-Caspase-8 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3020-100EA 8740 CZK
BIOV3020-100
Anti-Caspase-8 Rabbit Polyclonal Antibody
Caspase family of cysteine proteases has been shown to play a key role in apoptosis. Caspase-8 is a 55 kDa cytosolic protein that is synthesized as an inactive pro-enzyme. Activation of caspase-8 involves a two-step proteolysis: the cleavage of caspase-8 to generate a 43 and a 12 kDa fragment which is further processed to 10 kDa. The p43 is then cleaved to yield p26 and the release of the active site containing p18.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Caspase-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Bovine, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR