Tisk…

Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: IE6]

Dodavatel: BioVision
BIOV3022-100EA 6910 CZK
BIOV3022-100
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: IE6]
Protilátky
The DYKDDDDK Tag Antibody is a high-affinity mouse monoclonal antibody that can be used to detect DYKDDDDK-tagged proteins. It can detect N-terminal, C-terminal and internal tagged fusion proteins. This antibody recognizes the same epitope recognized by Sigma’s anti-FLAG® antibodies.

Recommended Dilutions: Western Blot: 0.5 µg/ml).

Type: Primary
Antigen: DYKDDDDK Tag
Clonality: Monoclonal
Clone: IE6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR