Tisk…

Anti-Cytochrome c Mouse Monoclonal Antibody [clone: 7H8.2C12]

Dodavatel: BioVision
BIOV3026-50EA 9590 CZK
BIOV3026-50
Anti-Cytochrome c Mouse Monoclonal Antibody [clone: 7H8.2C12]
Protilátky
Cytochrome c (m.w. 12,500) is an electron transport protein from mitochondria. It is released from mitochondria to cytoplasm during the early stages of apoptosis, prior to caspase activation, DNA fragmentation, and loss of membrane potential. The cytoplases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Cytochrome c
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR