Tisk…

Anti-Caspase-5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3029-100EA 7780 CZK
BIOV3029-100
Anti-Caspase-5 Rabbit Polyclonal Antibody
Caspase family of cysteine proteases has been shown to play a key role in apoptosis. Caspase-5 is also known as Ich-3 and ICErel-III and can proteolytically processed by granzyme B. Over expression of caspase-5 induces apoptosis in Rat-1 and HeLa cells, which can be inhibited by CrmA and Bcl-2.

Recommended Dilutions: Western Blot: 0.5-4ag/ml

Type: Primary
Antigen: Caspase-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR