Tisk…

Anti-BAK1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3031-100EA 6910 CZK
BIOV3031-100
Anti-BAK1 Rabbit Polyclonal Antibody
Protilátky
Bak is a homolog of apoptosis inhibitor Bcl-2. Bak is a member of the Bcl-2 family of proteins and contains Bcl-2 homology region (BH-) 1, 2 and 3. Unlike Bcl-2, Bak promotes cell death and counteracts the anti-apoptotic effect of Bcl-2. The immunoaffinity purified antibody detects a 30 kDa band, corresponding to the expected molecular weight of Bak on SDS-PAGE immunoblots.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunoprecipitation: 5-20 µg/ml).

Type: Primary
Antigen: BAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR