Tisk…

Anti-Bcl-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3033-100EA 8180 CZK
BIOV3033-100
Anti-Bcl-2 Rabbit Polyclonal Antibody
Protilátky
Bcl-2 and its homologs belong to a family of proteins that either promotes or prevent apoptosis. Bcl-2 alpha and beta are the alternatively spliced isoforms of 25 kDa and 22 kDa mitochondrial integral membrane proteins that inhibit apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipation: 4-8 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Bcl-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR