Tisk…

Anti-FADD Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3039-100EA 7760 CZK
BIOV3039-100
Anti-FADD Rabbit Polyclonal Antibody
Protilátky
Fas-associating death domain containing protein (FADD), also known as mediator of receptor induced toxicity (MORT1), is a 23 kDa cytosolic protein with an apparent molecular weight of 28 kDa on SDS-PAGE. The protein is involved in both Fas and TNF-R induced cell death pathway. FADD specifically binds to Fas to initiate the apoptotic signalling cascade. Overexpression of FADD induces apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; can also be used in immunoprecipitation

Type: Primary
Antigen: FADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR