Tisk…

Anti-PBR Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3044-100EA 8460 CZK
BIOV3044-100
Anti-PBR Rabbit Polyclonal Antibody
Protilátky
PBR (Peripheral-type Benzodiazepine Receptor) is a ubiquitous protein (18 kDa) found in most steroidogenic tissues. It is located primarily in the outer mitochondrial membrane of steroid-producing tissues and is found in association with a 34 kDa voltage-dependent anion channel protein (VDAC;PBR is involved in the regulation of cholesterol transport from the outer to the inner mitochondrial membrane. It is thoμght to be part of the mitochondrial permeability transition pore (PT) and to be involved in apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: PBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR