Tisk…

Anti-ASK1/MAPKKK5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3128-100EA 8430 CZK
BIOV3128-100
Anti-ASK1/MAPKKK5 Rabbit Polyclonal Antibody
ASK1 (Apoptosis Signal-regulating Kinase 1) activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activate c-Jun N-terminal Kinase (JNK) and p38 MAP Kinase. ASK1 is activated by TNFR and Fas throμgh the interaction with members of the TRAF Family and the Fas-associated protein Daxx. Overexpression of ASK1 induces apoptosis. However, a catalytically inactive form of ASK1 has been shown to inhibit TNF-α induced apoptosis.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ASK1/MAPKKK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR