Tisk…

Anti-LSD1/AOF2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3129-100EA 7780 CZK
BIOV3129-100
Anti-LSD1/AOF2 Rabbit Polyclonal Antibody
Lysine-specific demethylase 1(LSD1) is also known as AOF2, is a nuclear amine oxidase. LSD1 contain a Swirm domain, a FAD-binding motif and an amine oxidase domain that can act as a histone demethylase as well as transcription cofactor. LSD1 activity is inhibited by the amine oxidase inhibitors pargyline, deprenyl, clorgyline and tranylcypromine.

Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: LSD1/AOF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR