Tisk…

Anti-ABAD/HADH2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3246-100EA 7780 CZK
BIOV3246-100
Anti-ABAD/HADH2 Rabbit Polyclonal Antibody
HADH (Hydroxyacyl-Coenzyme A dehydrogenase) is a mitochondrial protein that catalyzes the oxidation of a wide variety of fatty acids, alcohols, and steroids. 3-hydroxyacyl-CoA dehydrogenase type II (HADH2) is a member of the short-chain dehydrogenase/reductase superfamily. HADH 2 is expressed at high levels in Alzheimer’s disease.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ABAD/HADH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR