Tisk…

Anti-Akt/PKB Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3247-100EA 9080 CZK
BIOV3247-100
Anti-Akt/PKB Rabbit Polyclonal Antibody
Akt, also referred to as PKB and RAC, is the cellular homologue of the viral oncogene v-Akt. This protein kinase is activated by insulin and various growth factors and functions in a pathway involving PI3 kinase as activation is sensitive to wortmanin. Recent evidence sμggests that Akt functions to promote cell survival by actively inhibiting apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-40 µg/ml; and Immunohistochemistry: 10-40 µg/ml

Type: Primary
Antigen: Akt/PKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR