Tisk…

Anti-IkBalpha Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3252-100EA 7780 CZK
BIOV3252-100
Anti-IkBalpha Rabbit Polyclonal Antibody
IkB-alpha protein is a NF-kB inhibitory protein. It binds NF-kB and retains the complex in the cytoplasm, thereby preventing it from entering into nucleus and functioning as a transcription factor. Upon receiving a variety of signals, IkB-alpha undergoes phosphorylation at serine residues by an ubiquitin-dependent protein kinases and finally degraded by the proteasome. Free NF-kB then binds DNA and affects gene expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IkBalpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR