Tisk…

Anti-ACAT2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3254-100EA 6910 CZK
BIOV3254-100
Anti-ACAT2 Rabbit Polyclonal Antibody
Protilátky
Acetyl-Coenzyme A acetyltransferase 2 (ACAT2) is an enzyme involved in lipid metabolism. Patients with ACAT2 deficiency have shown severe mental retardation and hypotonus. The ACAT2 gene shows complementary overlapping with the 3 prime region of the TCP1 gene in both mouse and human. These genes are encoded on opposite strands of DNA, as well as in opposite transcriptional orientation.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR