Tisk…

Anti-DAP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3261-100EA 9080 CZK
BIOV3261-100
Anti-DAP1 Rabbit Polyclonal Antibody
Death Associated Protein 1 (DAP1) is a 15 kDa protein that functions as a positive mediator of cell death initiated by interferon-gamma. The DAP1 protein is praline-rich and possesses one SH3 binding motif, as well as several consensus protein kinase phosphorylation sites. The protein is localized in the cytoplasma, however the detailed mechanism of its proapoptotic function is unclear.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR