Tisk…

Anti-alpha-Fodrin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3266-100EA 7780 CZK
BIOV3266-100
Anti-alpha-Fodrin Rabbit Polyclonal Antibody
Fodrin is a universally expressed membrane-associated cytoskeletal protein consisting of α- and β-subunits. The protein is important for maintaining normal membrane structure and supporting cell surface protein function. α-Fodrin is one of the important targets cleaved by caspases during apoptosis. The full length 240 kDa protein can be cleaved at several sites within its sequence by activated caspases generating N-terminal 150 kDa, C-terminal 120 kDa, and 35 kDa major products. Cleavage of α-Fodrin leads to membrane malfunction and cell shrinkage.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml;

Type: Primary
Antigen: alpha-Fodrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR