Tisk…

Anti-Lamin A Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3267-100EA 6910 CZK
BIOV3267-100
Anti-Lamin A Rabbit Polyclonal Antibody
Protilátky
Lamins are nuclear membrane structural components that are important in maintaining normal cell functions such as cell cycle control, DNA replication and chromatin organization. Lamin A is specifically cleaved by caspase-6 and therefore serves as a marker for caspase-6 activation. During apoptosis the 70 kDa lamin A is cleaved to a large (40-45 kDa) and small (23 kDa) fragment. The cleavage of lamins results in nuclear disregulation and cell death.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 15-20 µg/ml

Type: Primary
Antigen: Lamin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR