Tisk…

Anti-FATP4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3268-100EA 7780 CZK
BIOV3268-100
Anti-FATP4 Rabbit Polyclonal Antibody
FATP4 is a transmembrane protein that is involved in the uptake and metabolism of long chain fatty acids. FATP4 has acyl-CoA synthase activity and contains an AMP-binding motif. FATP4 is linked to insulin resistance and metabolic disorders.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: FATP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR