Tisk…

Anti-c-FLIP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3058-100EA 6910 CZK
BIOV3058-100
Anti-c-FLIP Rabbit Polyclonal Antibody
Protilátky
c-FLIP was cloned by several labs independently and termed Casper, I-FLICE, FLAME-1, CASH AND CLARP. c-FLIP contains two death effector domains and has sequence homology to caspase-8. c-FLIP interacts with adapter protein FADD and caspaser-8 and -10, and potently inhibits apoptosis induced by all known death receptors. Two splice variants of c-FLIP have been identified and termed FLIPl and FLIPs

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunofluorescence

Type: Primary
Antigen: c-FLIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR