Tisk…

Anti-PRMT7 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3059-100EA 6910 CZK
BIOV3059-100
Anti-PRMT7 Rabbit Polyclonal Antibody
Protilátky
Arginine methylation is an important protein modification catalyzed by arginine methyltransferase such as PRMT7. PRMT7 has two methyltransferase domains each containing a putative AdoMet binding motif. Arginine methylation has been implicated in signal transduction RNA processing and splicing.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PRMT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR