Tisk…

Anti-cdc42/Rac Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3080-100EA 8430 CZK
BIOV3080-100
Anti-cdc42/Rac Rabbit Polyclonal Antibody
cdc42/Rac belongs to the superfamily of small GTPases that are structurally linked to the proto-oncogene product p21ras and are important for the control of cell growth and differentiation as well as for intracellular organization. Cdc42/Rac is an important upstream regulator of the protein kinase cascade that controls the SAPK/JNK and p38 activity. Recent data also sμggest that constitutive active forms of cdc42 can induce apoptosis throμgh a mechanism requiring signaling throμgh SAPK/JNK.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: cdc42/Rac
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR