Tisk…
Anti-TCTN1 Rabbit Polyclonal Antibody
Dodavatel: BioVision
BIOV3087-100EA 7780 CZK
BIOV3087-100
Anti-TCTN1 Rabbit Polyclonal Antibody
TCTN1 belongs to the Tectonic protein family that regulate Hedgehog (Hh) pathway. During neural tube development, TCTN1 is required for the formation of most ventral cell types and for full Hh pathway activation. At later stages in development, TCTN1 is thoμght to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
Recommended Dilutions: Western Blot: 0.5-4 µg/ml
Type: Primary
Antigen: TCTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat