Tisk…

Anti-Lin28 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3091-200EA 6910 CZK
BIOV3091-200
Anti-Lin28 Rabbit Polyclonal Antibody
Protilátky
LIN28 is a highly conserved cytoplasmic RNA­binding protein. Lin28 has an N-terminal cold shock domain and two C­terminal (CCHC) zinc finger motif. It is expressed in embryonic stem cells and embryonic carcinoma cells. Lin28 is also found in adult cardiac and skeletal muscle.

Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: Lin28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR