Tisk…

Anti-BRD8 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3506-100EA 8430 CZK
BIOV3506-100
Anti-BRD8 Rabbit Polyclonal Antibody
Proinsulin is a prohormone precursor to Insulin, made by the pancreas in response to increased blood sμgar. Proinsulin is 39 amino acids longer than the mature Insulin, of which 4 are completely lost and the remaining 35 forms the C-peptide. The measurement of Proinsulin in serum provides a useful, valuable information for the diagnosis of insulinomas. Proinsulin levels have also been shown to be elevated in non-insulin dependent diabetics (NIDDM), in insulin dependent diabetics (IDDM) and other clinical situations. The antibody recognizes insulin and recombinant proinsulin, but does not react with synthetic C-peptide.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: BRD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR