Tisk…

Anti-NHERF1/ EBP50 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3521-100EA 7780 CZK
BIOV3521-100
Anti-NHERF1/ EBP50 Rabbit Polyclonal Antibody
NHE3 regulatory factor 1 (NHERF-1) is a multifunctional adaptor protein that interacts with NHE3 via its PDZ domains. The amino-terminal PDZ domain regulates the dimerization of NHERF-1 in vivo. G protein-coupled receptor kinase 6A phosphorylates NHERF-1 at Ser 289 via a PDZ domain-mediated interaction.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: NHERF1/ EBP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR