Tisk…

Anti-MMP-3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3523R-100EA 8120 CZK
BIOV3523R-100
Anti-MMP-3 Rabbit Polyclonal Antibody
Proteins of the matrix metalloprotease (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling as well as disease processes such as arthritis and metastasis. Most MMP’s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MMP-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Horse, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR