Tisk…

Anti-Caspase-6 (Active) Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3156-100EA 9590 CZK
BIOV3156-100
Anti-Caspase-6 (Active) Rabbit Polyclonal Antibody
Protilátky
Caspase family of cysteine proteases has been shown to play a key role in apoptosis. Similar to other caspases, caspase-6 is also synthesized as an inactive pro-enzyme that is processed in cells undergoing apoptosis. Together with caspase-3, caspase-6 is one of the major caspases in apoptotic cells, and functions downstream of apoptosis inhibitors Bcl-2 and Bcl-xL. Caspase-6 has also been shown involving in the proteolysis of poly (ADP-ribose) polymerase (PARP) and nuclear lamin A.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunofluorescence

Type: Primary
Antigen: Caspase-6 (Active)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR