Tisk…

Anti-BLK Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3175-100EA 8040 CZK
BIOV3175-100
Anti-BLK Rabbit Polyclonal Antibody
Protilátky
BLK, a BH3-containing protein that interacts with Bcl-2 and Bcl-xL, is a potent death agonist.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR