Tisk…

Anti-cIAP-1/HIAP-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3180-100EA 7900 CZK
BIOV3180-100
Anti-cIAP-1/HIAP-2 Rabbit Polyclonal Antibody
Protilátky
Apoptosis can be inhibited by a group of proteins called inhibitors of apoptosis (IAPs;These proteins contain a BIR (baculovirus IAP repeat) domain near the amino-terminus. The BIR domain can bind some caspases. Many members of the IAP family of proteins block proteolytic activation of caspase-3 and –7. For example, XIAP, cIAP-1 and cIAP-2 appear to block cytochrome c-induced activation of caspase-9, thereby preventing initiation of the caspase cascade. Since cIAP-1 and cIAP-2 were first identified as components in the cytosolic death domain-induced complex associated with the TNF family of receptors, they may inhibit apoptosis by additional mechanisms.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: cIAP-1/HIAP-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR