Tisk…

Anti-XIAP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3183-100EA 8120 CZK
BIOV3183-100
Anti-XIAP Rabbit Polyclonal Antibody
BIRC-4 (for baculoviral IAP repeat-containing protein 4) is a member of the IAP (for Inhibitor of apoptosis protein) family proteins. Other Aliases include RP1-315G1.5, API3, ILP1, MIHA, XIAP, XLP2. BIRC4 inhibits activated caspase-3 and apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in immunoprecipitation and Immunohistochemistry

Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR