Tisk…

Anti-MMP-13 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3533-100EA 8880 CZK
BIOV3533-100
Anti-MMP-13 Rabbit Polyclonal Antibody
Protilátky
The matrix metalloproteinases (MMP) are a family of peptidase enzymes responsible for the degradation of extracellular matrix components, including collagen, gelatin, fibronectin, laminin and proteoglycan. Transcription of MMP genes is differentially activated by phorbol ester, lipopolysaccharide (LPS) or staphylococcal enterotoxin B (SEB;MMP catalysis requires both calcium and zinc. MMP-13 (also designated collagenase-3) is produced by breast carcinomas and degrades collagen type I, II, III.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunoprecipitation and Immunohistochemistry: works with paraffin imbedded sections

Type: Primary
Antigen: MMP-13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Horse, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR