Tisk…

Anti-Fbx32/MAFbx/ATR-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3535-100EA 7190 CZK
BIOV3535-100
Anti-Fbx32/MAFbx/ATR-1 Rabbit Polyclonal Antibody
Protilátky
Atrogin-1 (Atr1) is also known as Muscle Atrophy F-box (MAFbx;It is an E3 ubiquitin ligase that initiates ATPdependent ubiquitin-mediated proteolysis and promotes muscle atrophy. MAFbx is approximately a 43kDa protein.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Fbx32/MAFbx/ATR-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR