Tisk…

Anti-MMP-17 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3537-100EA 7470 CZK
BIOV3537-100
Anti-MMP-17 Rabbit Polyclonal Antibody
Protilátky
The matrix metalloproteinases (MMP) are a family of peptidase enzymes responsible for the degradation of extracellular matrix components, including collagen, gelatin, fibronectin, laminin and proteoglycan. MMP-17 (also designated MT-MMP4 or MT4-MMP) is a glycosylphosphatidylinositol (GPI)-anchored proteinase and is mainly expressed in the brain, leukocytes, colon, ovary and testis.

Recommended Dilutions: Western Blot: 1-4 µg/ml; immunoprecipitation, and Immunohistochemistry

Type: Primary
Antigen: MMP-17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR