Tisk…

Anti-ACAA2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3538-100EA 6910 CZK
BIOV3538-100
Anti-ACAA2 Rabbit Polyclonal Antibody
Protilátky
ACAA2 (acetylcoenzyme A acyltransferase 2), is member of the thiolase family of enzymes and is involved in lipid metabolism. ACAA2 is localized in the mitochondria. ACCA2 catalyzes the last conversion step of acetyl-CoA to 3-oxoacyl-CoA, in the fatty acid oxidation pathway.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR