Tisk…

Anti-TLR5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3555R-100EA 7780 CZK
BIOV3555R-100
Anti-TLR5 Rabbit Polyclonal Antibody
The Toll-like receptor (TLR) family of proteins are characterized by a highly conserved Toll homology (TH) domain, which is essential for Toll-induced signal transduction. TLR1, as well as the other TLR family members, are type I transmembrane receptors that characteristically contain an extracellular domain consisting of several leucine-rich regions along with a single cytoplasmic Toll/IL-1R-like domain. TLR2 and TLR4 are activated in response to lipopolysacchride (LPS) stimulation, which results in the activation and translocation of NFkB and sμggests that these receptors are involved in mediating inflammatory responses. TLR6 is highly homologous to TLR1, and like other members of TLR family, it induces NFkB signaling upon activation. Expression of TLR receptors is highest in peripheral blood leukocytes, macrophages, and monocytes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR