Tisk…

Anti-TreakR Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3564-100EA 7470 CZK
BIOV3564-100
Anti-TreakR Rabbit Polyclonal Antibody
Protilátky
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrates that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by biding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in aorta and liver. It may also play a role in vascular cell migration and angiogenesis.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: TreakR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR