Tisk…

Anti-cAMP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3567-100EA 7780 CZK
BIOV3567-100
Anti-cAMP Rabbit Polyclonal Antibody
Affinity purified rabbit IgG to 2’O-succinyl adenosine-3’,5’ cyclic monophosphate (cyclic AMP, cAMP).

Recommended Dilutions: 1:50 dilutions for nonradioactive EIA assays (10 µl/assay in an 70 µl assay reaction;The material is sufficient for performing 500 nonradioactive EIA assays or 6,000 radioactive assays. Detect 1-100 fmol/assay in a 70 µl assay reaction using nonradioactive method. Refer to the cAMP Assay Kit (K371-100) for detailed assay protocol.

Type: Primary
Antigen: cAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR