Tisk…

Anti-Phospho-IRS (Ser616) Mouse Monoclonal Antibody [clone: HIR-B1]

Dodavatel: BioVision
BIOV3105-100EA 9080 CZK
BIOV3105-100
Anti-Phospho-IRS (Ser616) Mouse Monoclonal Antibody [clone: HIR-B1]
Insulin receptor substrates (IRS) subtypes are responsible for several insulin related activities, such as glucose homeostasis, cell growth, cell transformation, apoptosis and insulin signal transduction. The balance between Ser/Thr phosphorylation of IRS has been demonstrated to be an important regulator of insulin signaling. IRS1 has also been shown to be constitutively activated in cancers such as breast cancer, Wilm’s tumors, and adrenal cortical carcinomas. Does not recognize IRS peptide NOT phosphorylated at Ser616 and will not recognize IRS peptide phosphorylated at Tyr183. Also does not react with riboflavin carrier protein which is known to have multiple phosphate groups.

Recommended Dilutions: indirect ELISA and WB.

Type: Primary
Antigen: Phospho-IRS (Ser616)
Clonality: Monoclonal
Clone: HIR-B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR