Tisk…

Anti-p38 MAP Kinase Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3114-100EA 8120 CZK
BIOV3114-100
Anti-p38 MAP Kinase Rabbit Polyclonal Antibody
p38 MAP kinase is the mammalian homologue of the yeast HOG kinase and participates in a cascade controlling cell cellular responses to cytokines and stress. Like the SAPK/JNK pathway, p38 MAP kinase is activated by a variety of cellular stresses including inflammatory cytokines, UV light and growth factors, etc. Activated p38 MAP kinase has been shown to phosphorylate and activate MAPKAP kinase-2 and to phosphorylate the transcription factors ATF-2 and Max.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: p38 MAP Kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR