Tisk…

Anti-PKA Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3115-100EA 6910 CZK
BIOV3115-100
Anti-PKA Rabbit Polyclonal Antibody
Protilátky
Protein kinase A (PKA) also known as cAMP-dependent protein kinase, is a 40 kDa protein kinase that phosphorylates serine or threonine residues in target proteins in response to elevated levels of cAMP. It has been sμggested that the complex structure of PKA may be necessary for it to accomplish such diverse functions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 20-30 µg/ml

Type: Primary
Antigen: PKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR